Trisodium-N-(2-hydroxyethyl)ethylenediamine-N,N',N'-triacetate dihydrate ≥98.0% (by titrimetric analysis)
Fornitore: TCI
|
Attenzione
|
Formula:
C₁₀H₁₅N₂Na₃O₇·2H₂O Punto di fusione: 288°C |
MDL Number:
MFCD00043341 CAS Number: 139-89-9 EINECS: 205-381-9 Merck Index: 14,09966 |
|
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (Chelometric Titration) | min. 98.0 % (calcd.on anh.substance) |
Water | 5.0 to 12.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...