Stampa…

Harmine hydrochloride ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: Banisterine Hydrochloride

H0002-1G H0002-100MG
TCIAH0002-1GEA 94 EUR
TCIAH0002-1G TCIAH0002-100MG
Harmine hydrochloride ≥98.0% (by HPLC, titration analysis)
Harmine hydrochloride
Formula: C₁₃H₁₂N₂O
Peso molecolare: 248,71 g/mol
MDL Number: MFCD00012641
CAS Number: 343-27-1
EINECS: 206-443-8
UN: 1544
ADR: 6.1,III
Merck Index: 14,04616

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 % (calcd.on dried substance)
Water max. 8.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR