4-Glycidyloxy-2,2,6,6-tetramethylpiperidine 1-oxyl ≥95.0% (by GC, titration analysis) free radical
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
4-Glycidyloxy-TEMPO Free Radical
G0555-1G
G0555-5G
TCIAG0555-1GEA
53
EUR
TCIAG0555-1G
TCIAG0555-5G
4-Glycidyloxy-2,2,6,6-tetramethylpiperidine 1-oxyl ≥95.0% (by GC, titration analysis) free radical
4-Glycidyloxy-2,2,6,6-tetramethylpiperidine 1-oxyl
Formula:
C₁₂H₂₂NO₃ Peso molecolare: 228,31 g/mol Punto di fusione: 38°C Temperatura di stoccaggio: Refrigerata |
CAS Number:
122413-85-8 |
Risultati del test di specificazione
Appearance | Orange to Brown powder to crystal |
Purity (GC) | min. 95.0 % |
Purity (Iodometric Titration) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...