Stampa…

Gomisin A ≥96.0% (by HPLC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Besigomsin, Schisandrol B, (6S,7S,13aR)-5,6,7,8-Tetrahydro-1,2,3,13-tetramethoxy-6,7-dimethylbenzo[3,4]cycloocta[1,2-f][1,3]benzodioxol-6-ol, (6S,7S)-1,2,3,13-Tetramethoxy-6,7-dimethyl-5,6,7,8-tetrahydrobenzo[3',4']cycloocta[1',2':4,5]benzo[1,2-d][1,3]dioxol-6-ol, (6S,7S,13aR)-1,2,3,13-Tetramethoxy-6,7-dimethyl-5,6,7,8-tetrahydrobenzo[3',4']cycloocta[1',2':4,5]benzo[1,2-d][1,3]dioxol-6-ol

G0537-25MG G0537-5MG
TCIAG0537-25MGEA 238 EUR
TCIAG0537-25MG TCIAG0537-5MG
Gomisin A ≥96.0% (by HPLC)
Gomisin A
Formula: C₂₃H₂₈O₇
Peso molecolare: 416,47 g/mol
Temperatura di stoccaggio: Congelamento
MDL Number: MFCD01941645
CAS Number: 58546-54-6

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 96.0 area%
Melting point 91.0 to 95.0 deg-C
Specific rotation [a]20/D +67.0 to +73.0 deg (C=1, CHCl3)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR