Germacrone ≥98.0% (by GC)
Fornitore: TCI
|
Attenzione
|
G0519-20MG
G0519-100MG
TCIAG0519-20MGEA
127
EUR
TCIAG0519-20MG
TCIAG0519-100MG
Germacrone ≥98.0% (by GC)
Germacrone
Formula:
C₁₅H₂₂O Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00210050 CAS Number: 6902-91-6 |
|
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 54.0 to 58.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...