Stampa…

Glyceryl Ascorbate ≥95.0% (by titrimetric analysis)

Fornitore: TCI EUROPE

Synonyms: 2-O-(2,3-Dihydroxypropyl)-L-ascorbic acid

G0451-5G G0451-25G
TCIAG0451-5GEA 55 EUR
TCIAG0451-5G TCIAG0451-25G
Glyceryl Ascorbate ≥95.0% (by titrimetric analysis)
Glyceryl Ascorbate
Formula: C₉H₁₄O₈
Peso molecolare: 250,20 g/mol
CAS Number: 1120360-13-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=019f9d45f3dbb076d96c68e35c6a517d4dcdad36&vid=019f9d45f3dbb076d96c68e35c6a517d4dcdad36&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 95.0 %
Melting point 156.0 to 161.0 deg-C
Specific rotation [a]20/D +52.0 to +57.0 deg (C=1, MeOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR