Guanidine hydroiodide ≥97.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
G0450-1G
G0450-5G
TCIAG0450-1GEA
32
EUR
TCIAG0450-1G
TCIAG0450-5G
Guanidine hydroiodide ≥97.0% (by titrimetric analysis)
Guanidine hydroiodide
Formula:
CH₅N₃ Peso molecolare: 186,98 g/mol |
CAS Number:
19227-70-4 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (Nonaqueous Titration) | min. 97.0 % |
Purity (Argentmetric Titration) | min. 97.0 % |
Melting point | 194.0 to 198.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...