Glycidyl lauryl ether ≥97.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
Dodecyl Glycidyl Ether
G0448-1G
G0448-5G
TCIAG0448-1GEA
41
EUR
TCIAG0448-1G
TCIAG0448-5G
Glycidyl lauryl ether ≥97.0% (by GC)
Glycidyl lauryl ether
Formula:
C₁₅H₃₀O₂ Peso molecolare: 242,40 g/mol Punto di ebollizione: 324°C |
MDL Number:
MFCD00022344 CAS Number: 2461-18-9 EINECS: 219-554-1 |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...