Guanfacine hydrochloride ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
N-Amidino-2-(2,6-dichlorophenyl)acetamide Hydrochloride, N-Carbamimidoyl-2-(2,6-dichlorophenyl)acetamide hydrochloride, N-(Aminoiminomethyl)-2,6-dichlorobenzeneacetamide Hydrochloride
G0414-100MG
G0414-1G
TCIAG0414-100MGEA
39
EUR
TCIAG0414-100MG
TCIAG0414-1G
Guanfacine hydrochloride ≥98.0% (by HPLC, titration analysis)
Guanfacine hydrochloride
Formula:
C₉H₉Cl₂N₃O Peso molecolare: 282,55 g/mol |
MDL Number:
MFCD00798230 CAS Number: 29110-48-3 EINECS: 249-443-3 UN: 2811 ADR: 6.1,III Merck Index: 14,04561 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 211.0 to 215.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...