Stampa…

Glipizide ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI

G0369-1G G0369-5G
TCIAG0369-1GEA 69 EUR
TCIAG0369-1G TCIAG0369-5G
Glipizide ≥98.0% (by HPLC, titration analysis)
Glipizide
Formula: C₂₁H₂₇N₅O₄S
MDL Number: MFCD00072159
CAS Number: 29094-61-9
EINECS: 249-427-6
Merck Index: 14,04442

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR