Stampa…

Gemcitabine hydrochloride ≥98.0% (by HPLC, total nitrogen)

Fornitore: TCI
Attenzione

Synonyms: 2'-Deoxy-2',2'-difluorocytidine, dFdC, 4-Amino-1-((2R,4R,5R)-3,3-difluoro-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one hydrochloride

G0367-1G G0367-100MG
TCIAG0367-1GEA 39 EUR
TCIAG0367-1G TCIAG0367-100MG
Gemcitabine hydrochloride ≥98.0% (by HPLC, total nitrogen)
Gemcitabine hydrochloride
Formula: C₉H₁₂ClF₂N₃O₄
Peso molecolare: 299.6591 g/mol
Punto di fusione: 292°C
MDL Number: MFCD01735988
CAS Number: 122111-03-9
Merck Index: 14,04386

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Specific rotation [a]20/D +46.0 to +49.0 deg (C=1, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR