Stampa…

(R)-N-Glycidylphthalimide ≥99.0% (by GC)

Fornitore: TCI EUROPE
Pericolo

Synonyms: (R)-N-(2,3-Epoxypropyl)phthalimide, (R)-2-(Oxiran-2-ylmethyl)isoindoline-1,3-dione

G0327-25G G0327-5G
TCIAG0327-25GEA 191 EUR
TCIAG0327-25G TCIAG0327-5G
(R)-N-Glycidylphthalimide ≥99.0% (by GC)
(R)-N-Glycidylphthalimide
Formula: C₁₁H₉NO₃
Peso molecolare: 203,20 g/mol
MDL Number: MFCD04973349
CAS Number: 181140-34-1

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 99.0 %
Melting point 99.0 to 103.0 deg-C
Specific rotation [a]20/D -7.0 to -13.0 deg (C=2.2, CHCl3)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR