Ganciclovir ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
9-[[2-Hydroxy-1-(hydroxymethyl)ethoxy]methyl]guanine, 2'-Nor-2'-Deoxyguanosine, BW 759, 2-Amino-9-(((1,3-dihydroxypropan-2-yl)oxy)methyl)-1H-purin-6(9H)-one, RS-21592
G0315-5G
G0315-25G
TCIAG0315-5GEA
33
EUR
TCIAG0315-5G
TCIAG0315-25G
Ganciclovir ≥98.0% (by HPLC, titration analysis)
Ganciclovir
Formula:
C₉H₁₃N₅O₄ Peso molecolare: 255,23 g/mol Punto di fusione: 250°C Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00870588 CAS Number: 82410-32-0 Merck Index: 14,04363 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % (calcd.on dried substance) |
Drying loss | max. 4.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...