Gal[2346Ac]β1-3)GlcN3[46Bzd]-β-MP 98,0 HPLC_ASSAY_METHOD
Fornitore: TCI
Synonyms:
4-Methoxyphenyl 3-O-(2,3,4,6-Tetra-O-acetyl-β-D-galactopyranosyl)-2-azido-4,6-O-benzylidene-2-deoxy-β-D-glucopyranoside
|
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Melting point | 170.0 to 174.0 deg-C |
Specific rotation [a]20/D | -12.0 to -15.0 deg (C=1, CHCl3) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...