GlcNPhth[346Ac]-β-(1-3)Gal[246Bn]-β-MP ≥98.0% (by HPLC)
Fornitore: TCI
Synonyms:
4-Methoxyphenyl 3-O-(3,4,6-Tri-O-acetyl-2-deoxy-2-phthalimido-β-D-glucopyranosyl)-2,4,6-tri-O-benzyl-β-D-galactopyranoside
Formula:
C₅₄H₅₅NO₁₆ Temperatura di stoccaggio: Refrigerata |
CAS Number:
1820575-44-7 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Melting point | 173.0 to 177.0 deg-C |
Specific rotation [a]20/D | -7.0 to -10.0 deg (C=1, CHCl3) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...