(S)-Glycidyl trityl ether ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
(S)-Glycidyl Triphenylmethyl Ether, (S)-2-(Triphenylmethoxymethyl)oxirane, (S)-2-((Trityloxy)methyl)oxirane
G0285-25G
G0285-5G
TCIAG0285-25GEA
193
EUR
TCIAG0285-25G
TCIAG0285-5G
(S)-Glycidyl trityl ether ≥98.0% (by GC)
(S)-Glycidyl trityl ether
Formula:
C₂₂H₂₀O₂ Peso molecolare: 316,40 g/mol |
MDL Number:
MFCD00273373 CAS Number: 129940-50-7 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Optical purity (LC) | min. 98.0 ee% |
Melting point | 99.0 to 103.0 deg-C |
Specific rotation [a]20/D | -10.0 to -12.0 deg (C=1, CHCl3) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...