3-Glycidyloxypropyl(dimethoxy)methylsilane ≥96.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
3-Glycidoxypropyldimethoxymethylsilane, 3-[Dimethoxy(methyl)silyl]propyl Glycidyl Ether
G0261-25G
G0261-500G
TCIAG0261-25GEA
36
EUR
TCIAG0261-25G
TCIAG0261-500G
3-Glycidyloxypropyl(dimethoxy)methylsilane ≥96.0% (by GC)
3-Glycidyloxypropyl(dimethoxy)methylsilane
Risultati del test di specificazione
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...