Stampa…

2-[2-(2-Azidoethoxy)ethoxy]ethyl-2,3,4,6-tetra-O-acetyl-D-galactopyranoside ≥95.0% (by total nitrogen basis)

Fornitore: TCI

Synonyms: 2-[2-(2-Azidoethoxy)ethoxy]ethyl 2,3,4,6-Tetra-O-acetyl-D-galactopyranoside

G0257-1G G0257-5G
TCIAG0257-1GEA 186 EUR
TCIAG0257-1G TCIAG0257-5G
2-[2-(2-Azidoethoxy)ethoxy]ethyl-2,3,4,6-tetra-O-acetyl-D-galactopyranoside ≥95.0% (by total nitrogen basis)
2-[2-(2-Azidoethoxy)ethoxy]ethyl-2,3,4,6-tetra-O-acetyl-D-galactopyranoside
Formula: C₂₀H₃₁N₃O₁₂
Peso molecolare: 505.48 g/mol
MDL Number: MFCD00191441
CAS Number: 381716-33-2

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white clear liquid to slightly cloudy liquid
Purity (with Total Nitrogen) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR