Penta-O-acetyl-β-D-galactopyranose ≥97.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
β-D-Galactose Pentaacetate, (2S,3R,4S,5S,6R)-6-(Acetoxymethyl)tetrahydro-2H-pyran-2,3,4,5-tetrayl tetraacetate
G0247-25G
G0247-250G
TCIAG0247-25GEA
33
EUR
TCIAG0247-25G
TCIAG0247-250G
Penta-O-acetyl-β-D-galactopyranose ≥97.0% (by GC)
Penta-O-acetyl-beta-D-galactopyranose
Formula:
C₁₆H₂₂O₁₁ Peso molecolare: 390,34 g/mol |
MDL Number:
MFCD00063259 CAS Number: 4163-60-4 EINECS: 224-008-0 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 143.0 to 146.0 deg-C |
Specific rotation [a]20/D | +21.0 to +26.0 deg (C=2, Methanol) |
Solubility in hot Methanol | very faint turbidity |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...