Stampa…

Penta-O-acetyl-ɑ-D-glucopyranose ≥97.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: 1,2,3,4,6-Penta-O-acetyl-α-D-glycopyranose, ɑ-D-Glucose Pentaacetate, 1,2,3,4,6-Penta-O-Acetyl-A-D-Glycopyranose, α-D(+)-Glucose pentaacetate, (2R,3R,4S,5R,6R)-6-(Acetoxymethyl)tetrahydro-2H-pyran-2,3,4,5-tetrayl tetraacetate, Pentaacetyl-ɑ-D-glucose, α-D-Glucose pentaacetate

G0225-25G G0225-250G
TCIAG0225-25GEA 18 EUR
TCIAG0225-25G TCIAG0225-250G
Penta-O-acetyl-ɑ-D-glucopyranose ≥97.0% (by GC)
Penta-O-acetyl-alpha-D-glucopyranose
Formula: C₁₆H₂₂O₁₁
Peso molecolare: 390,34 g/mol
MDL Number: MFCD00064071
CAS Number: 604-68-2
EINECS: 210-073-2

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Melting point 110.0 to 114.0 deg-C
Specific rotation [a]20/D +104 to +107 deg (C=1.5, MeOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR