Stampa…

Glycine zinc salt monohydrate ≥97.0% (by titrimetric analysis) for protein research

Fornitore: TCI

Synonyms: Zinc Glycinate

G0215-25G G0215-5G
TCIAG0215-25GEA 162 EUR
TCIAG0215-25G TCIAG0215-5G
Glycine zinc salt monohydrate ≥97.0% (by titrimetric analysis) for protein research
Glycine zinc salt monohydrate
Formula: C₂H₅NO₂
Peso molecolare: 213,50 g/mol
MDL Number: MFCD00070561
CAS Number: 14281-83-5
EINECS: 238-173-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5fb4c8bedc731f23c9bd6b81924c86ae2fc1af35&vid=5fb4c8bedc731f23c9bd6b81924c86ae2fc1af35&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Chelometric Titration) min. 97.0 % (calcd.on anh.substance)
Water 7.4 to 8.2 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR