Stampa…

Sodium glucoheptonate dihydrate ≥98.0% (by titrimetric analysis)

Fornitore: TCI

Synonyms: Glucosecarboxylic Acid Sodium Salt, Glucoheptonic Acid Sodium Salt

G0214-25G G0214-500G
TCIAG0214-25GEA 16 EUR
TCIAG0214-25G TCIAG0214-500G
Sodium glucoheptonate dihydrate ≥98.0% (by titrimetric analysis)
Sodium glucoheptonate dihydrate
Formula: C₇H₁₄O₈
Peso molecolare: 248.16 g/mol
MDL Number: MFCD00013425
CAS Number: 31138-65-5
EINECS: 250-480-2

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystaline
Purity (Nonaqueous Titration) min. 98.0 % (calcd.on anh.substance)
Specific rotation [a]20/D +5.0 to +6.0 deg (C=10, H2O)
Water 11.0 to 14.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR