Stampa…

Glyoxalbis(2-hydroxyanil) ≥98.0% (by total nitrogen basis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2,2'-Ethanediylidenedinitrilodiphenol, 2,2'-(Ethane-1,2-diylidenebis(azanylylidene))diphenol, Glyoxalbis(2-hydroxyanil), 1,2-Bis(2-hydroxyphenylimino)ethane

G0153-25G G0153-5G
TCIAG0153-25GEA 131 EUR
TCIAG0153-25G TCIAG0153-5G
Glyoxalbis(2-hydroxyanil) ≥98.0% (by total nitrogen basis)
Glyoxalbis(2-hydroxyanil)
Formula: C₁₄H₁₂N₂O₂
Peso molecolare: 240,26 g/mol
Punto di fusione: 205°C
MDL Number: MFCD00002183
CAS Number: 1149-16-2
EINECS: 214-560-0

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Solubility in EtOH almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR