Glycyrrhizin ≥93.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
Synonyms:
Glycyrrhizic Acid
G0150-25G
G0150-1G
TCIAG0150-25GEA
93
EUR
TCIAG0150-25G
TCIAG0150-1G
Glycyrrhizin ≥93.0% (by titrimetric analysis)
Glycyrrhizin
Formula:
C₄₂H₆₂O₁₆ Peso molecolare: 822,94 g/mol |
MDL Number:
MFCD00065194 CAS Number: 1405-86-3 EINECS: 215-785-7 Merck Index: 14,04505 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 75.0 area% |
Purity (Neutralization titration) | min. 93.0 % (calcd.on anh.substance) |
Specific rotation [a]20/D | +56.0 to +63.0 deg (C=1.5, EtOH) (calcd.on anh.substance) |
Water | max. 7.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...