Stampa…

Glycyl-l-valine

Fornitore: TCI

G0148-1G G0148-5G
TCIAG0148-1GEA 48 EUR
TCIAG0148-1G TCIAG0148-5G
Glycyl-l-valine
Proteine e Peptidi

  • Specific rotation [a]20/D: –20° (C=2, H₂O)
  • Solubility in water: Almost transparency
  • EINECS: 217-806-5


Product Categories: Bioscience, Oligopeptides
Other Product Names / Synonyms: H-Gly-Val-OH

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR