Stampa…

Glycyl-DL-norleucine ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE

Synonyms: H-Gly-DL-Nle-OH

G0133-1G G0133-5G
TCIAG0133-1GEA 62 EUR
TCIAG0133-1G TCIAG0133-5G
Glycyl-DL-norleucine ≥98.0% (by titrimetric analysis)
Glycyl-DL-norleucine
Formula: C₈H₁₆N₂O₃
Peso molecolare: 188,23 g/mol
MDL Number: MFCD00038187
CAS Number: 1504-41-2

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 221.0 to 227.0 deg-C
Solubility in hot Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR