Glycy- l-alanine
Fornitore: TCI
G0113-1G
G0113-5G
TCIAG0113-1GEA
50
EUR
TCIAG0113-1G
TCIAG0113-5G
Glycy- l-alanine
Proteine e Peptidi
- Specific rotation [a]20/D: –48,0 to –51,0 °C (C=2, H₂O)
- Solubility in water: Almost transparency
- EINECS: 223-019-8
- Product Categories: Bioscience, Oligopeptides
- Other Product Names / Synonyms: H-Gly-Ala-OH
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...