Monoolein 40,00% (by GC)
Fornitore: TCI EUROPE
Synonyms:
9-Octadecenoicacid(Z)-,2,3-dihydroxypropylester, 1-(cis-9-Octadecenoyl)-rac-glycerol, Glyceryl cis-9-octadecenate hygroscopic, DL-α-Monoolein, rac-Glycerol 1-monooleate, 1-Oleoyl-rac-glycerol, 1-Mono(cis-9-octadecenoyl)-rac-glycerol, 1-Monleoyl-rac-glycerol, Glyceryl monooleate
Formula:
C₂₁H₄₀O₄ Peso molecolare: 356,547 g/mol Flash Pt: 180°C |
MDL Number:
MFCD00042735 CAS Number: 111-03-5 EINECS: 203-827-7 |
Risultati del test di specificazione
Appearance | White to Yellow to Orange clear liquid |
Purity (GC) | min. 40.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...