Stampa…

Diacetin (contains Mono-, Tri-, Glycerol)

Fornitore: TCI EUROPE

Synonyms: Glycerol alpha,alpha'-Diacetate, 2-Hydroxypropane-1,3-diyl diacetate

G0077-25G G0077-500G
TCIAG0077-25GEA 16 EUR
TCIAG0077-25G TCIAG0077-500G
Diacetin (contains Mono-, Tri-, Glycerol)
Diacetin
Formula: C₇H₁₂O₅
Peso molecolare: 176,17 g/mol
Punto di ebollizione: 261°C
Flash Pt: 146°C
MDL Number: MFCD00008717
CAS Number: 25395-31-7
EINECS: 246-941-2

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 40.0% (total of isomer)
Acid Value max. 3.0

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR