Stampa…

Disodium glutarate ≥99.0% (by titrimetric analysis)

Fornitore: TCI EUROPE

Synonyms: Glutaric Acid Disodium Salt

G0070-25G G0070-500G
TCIAG0070-25GEA 42 EUR
TCIAG0070-25G TCIAG0070-500G
Disodium glutarate ≥99.0% (by titrimetric analysis)
Disodium glutarate
Formula: C₅H₈O₄
Peso molecolare: 176,08 g/mol
MDL Number: MFCD00070497
CAS Number: 13521-83-0
EINECS: 236-864-2

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 99.0 %
Solubility in Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR