Stampa…

D-Glucose-6-phosphate barium salt heptahydrate ≥98.0% (W)

Fornitore: TCI EUROPE
Pericolo

Synonyms: D-Glucose-6-Phosphate Barium Salt Hydrate

G0052-1G G0052-100MG
TCIAG0052-1GEA 56 EUR
TCIAG0052-1G TCIAG0052-100MG
D-Glucose-6-phosphate barium salt heptahydrate ≥98.0% (W)
D-Glucose-6-phosphate barium salt heptahydrate
Formula: C₆H₁₃O₉P
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00221521
CAS Number: 58823-95-3
EINECS: 261-463-4
UN: 1564
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6d5e72a35eb35a40529f8e5c3564b344deb2d8a0&vid=6d5e72a35eb35a40529f8e5c3564b344deb2d8a0&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (Gravimetric Method) min. 98.0 % (calcd.on anh.substance)
Specific rotation [a]20/D +18.0 to +22.0 deg (C=5, 1mol/L HCl)
Water 20.0 to 25.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR