Stampa…

4-Fluoroaniline Hydroiodide ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 4-Fluoroanilinium iodide, 4-Fluorophenylammonium iodide, 4-Fluorophenylamine Hydroiodide

F1273-1G F1273-5G
TCIAF1273-1GEA 56 EUR
TCIAF1273-1G TCIAF1273-5G
4-Fluoroaniline Hydroiodide ≥98.0% (by HPLC, titration analysis)
4-Fluoroaniline Hydroiodide
Formula: C₆H₆FN·HI
Peso molecolare: 239,03 g/mol
CAS Number: 85734-19-6

Visualizza i codici

Risultati del test di specificazione

Appearance White to Gray to Red powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Argentmetric Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR