Stampa…

1-Fluorocyclopropanecarboxylic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Pericolo

F1254-200MG F1254-1G
TCIAF1254-200MGEA 40 EUR
TCIAF1254-200MG TCIAF1254-1G
1-Fluorocyclopropanecarboxylic acid ≥98.0% (by GC, titration analysis)
1-Fluorocyclopropanecarboxylic acid
Formula: C₄H₅FO₂
Peso molecolare: 104,08 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD18205954
CAS Number: 137081-41-5
UN: 3261
ADR: 8,II

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 72.0 to 76.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR