1,3,5-Tris(4-formylphenyl)benzene ≥96.0% (by HPLC)
Fornitore: TCI EUROPE
Synonyms:
5'-(4-Formylphenyl)-[1,1':3',1''-terphenyl]-4,4''-dicarboxaldehyde, 5'-(4-Formylphenyl)-[1,1':3',1''-terphenyl]-4,4''-dicarbaldehyde
F1252-200MG
F1252-1G
TCIAF1252-200MGEA
75
EUR
TCIAF1252-200MG
TCIAF1252-1G
1,3,5-Tris(4-formylphenyl)benzene ≥96.0% (by HPLC)
1,3,5-Tris(4-formylphenyl)benzene
Formula:
C₂₇H₁₈O₃ Peso molecolare: 390,44 g/mol |
MDL Number:
MFCD27937712 CAS Number: 118688-53-2 |
Risultati del test di specificazione
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (HPLC) | min. 96.0 area% |
Melting point | 248.0 to 252.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...