Stampa…

Formamidine hydrobromide ≥99.0% (by total nitrogen and titration analysis) for perovskite precursor

Fornitore: TCI EUROPE

Synonyms: Formimidamide hydrobromide, Formamidinium bromide, FABr

F1244-1G F1244-25G F1244-5G
TCIAF1244-1GEA 45 EUR
TCIAF1244-1G TCIAF1244-25G TCIAF1244-5G
Formamidine hydrobromide ≥99.0% (by total nitrogen and titration analysis) for perovskite precursor
Formamidine hydrobromide
Formula: CH₄N₂·HBr
Peso molecolare: 124,97 g/mol
MDL Number: MFCD28369274
CAS Number: 146958-06-7

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 99.0 %
Purity (Argentmetric Titration) min. 99.0 %
Melting point 133.0 to 137.0 deg-C
Elemental analysis (Carbon) 9.30 to 9.80 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR