(Ferrocenylmethyl)trimethylammonium chloride ≥98.0% (by HPLC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
(Trimethylammoniomethyl)ferrocene chloride, FcNCl
F1231-5G
F1231-1G
TCIAF1231-5GEA
240
EUR
TCIAF1231-5G
TCIAF1231-1G
(Ferrocenylmethyl)trimethylammonium chloride ≥98.0% (by HPLC)
(Ferrocenylmethyl)trimethylammonium chloride
Formula:
C₁₄H₂₀ClFeN Peso molecolare: 293,62 g/mol |
CAS Number:
83617-79-2 |
Risultati del test di specificazione
Appearance | Light yellow to Brown powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Argentmetric Titration) | min. 95.0 % |
Water | max. 5.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...