Stampa…

2-(4-Fluorophenyl)ethylamine hydrobromide ≥98.0% (by HPLC, total nitrogen)

Fornitore: TCI
Attenzione

F1229-1G F1229-5G
TCIAF1229-1GEA 58 EUR
TCIAF1229-1G TCIAF1229-5G
2-(4-Fluorophenyl)ethylamine hydrobromide ≥98.0% (by HPLC, total nitrogen)
2-(4-Fluorophenyl)ethylamine hydrobromide
Formula: C₈H₁₀FN·HBr
CAS Number: 1807536-06-6

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR