1-(Furan-2-yl)but-3-en-1-ol ≥95.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
alpha-2-Propen-1-yl-2-furanmethanol
F1226-5G
F1226-1G
TCIAF1226-5GEA
147
EUR
TCIAF1226-5G
TCIAF1226-1G
1-(Furan-2-yl)but-3-en-1-ol ≥95.0% (by GC)
1-(Furan-2-yl)but-3-en-1-ol
Formula:
C₈H₁₀O₂ Peso molecolare: 138,17 g/mol |
MDL Number:
MFCD03093935 CAS Number: 6398-51-2 |
Risultati del test di specificazione
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 95.0 % |
Specific gravity (20/20) | 1.0420 to 1.0520 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...