2-Fluoro-4,4'-bis(trans-4-propylcyclohexyl)biphenyl ≥98.0% (by GC)
Fornitore: TCI EUROPE
F1217-1G
F1217-5G
TCIAF1217-1GEA
68
EUR
TCIAF1217-1G
TCIAF1217-5G
2-Fluoro-4,4'-bis(trans-4-propylcyclohexyl)biphenyl ≥98.0% (by GC)
2-Fluoro-4,4'-bis(trans-4-propylcyclohexyl)biphenyl
Formula:
C₃₀H₄₁F Peso molecolare: 420,656 g/mol |
MDL Number:
MFCD11053361 CAS Number: 102714-93-2 EINECS: 441-120-7 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...