Stampa…

FR180204 ≥98.0% (by HPLC)

Fornitore: TCI EUROPE

Synonyms: FR180204, FR 180204, FR-180204, 5-(2-phenylpyrazolo[1,5-a]pyridin-3-yl)-2H-pyrazolo[3,4-c]pyridazin-3-amine, 5-(2-Phenylpyrazolo[1,5-a]pyridin-3-yl)-1H-pyrazolo[3,4-c]pyridazin-3-amine

F1214-25MG F1214-5MG
TCIAF1214-25MGEA 420 EUR
TCIAF1214-25MG TCIAF1214-5MG
FR180204 ≥98.0% (by HPLC)
FR180204
Formula: C₁₈H₁₃N₇
Peso molecolare: 327,34 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD14585805
CAS Number: 865362-74-9

Visualizza i codici

Risultati del test di specificazione

Appearance Light orange to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 28.50 to 31.50 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR