Stampa…

1-(4-Fluorophenyl)-3-(phenylsulphonyl)-1H-pyrrolo[2,3-b]quinoxalin-2-amine ≥95.0% (by HPLC)

Fornitore: TCI EUROPE

Synonyms: CAY10602, CAY 10602

F1204-10MG F1204-50MG
TCIAF1204-10MGEA 68 EUR
TCIAF1204-10MG TCIAF1204-50MG
1-(4-Fluorophenyl)-3-(phenylsulphonyl)-1H-pyrrolo[2,3-b]quinoxalin-2-amine ≥95.0% (by HPLC)
1-(4-Fluorophenyl)-3-(phenylsulphonyl)-1H-pyrrolo[2,3-b]quinoxalin-2-amine
Formula: C₂₂H₁₅FN₄O₂S
Peso molecolare: 418,45 g/mol
Temperatura di stoccaggio: Congelamento
MDL Number: MFCD02236691
CAS Number: 374922-43-7

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (HPLC) min. 95.0 area%
Elemental analysis (Nitrogen) 12.70 to 14.00 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR