Fmoc-D-Phg-OH ≥97.0% (by HPLC, titration analysis)
Fornitore: TCI EUROPE
Synonyms:
Fmoc-D-Phg-OH, N-Fmoc-D-α-phenylglycine, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-2-phenylglycine, N-Fmoc-D-2-phenylglycine
F1193-25G
F1193-5G
TCIAF1193-25GEA
106
EUR
TCIAF1193-25G
TCIAF1193-5G
Fmoc-D-Phg-OH ≥97.0% (by HPLC, titration analysis)
Fmoc-D-Phg-OH
Formula:
C₂₃H₁₉NO₄ Peso molecolare: 373,408 g/mol |
MDL Number:
MFCD00190891 CAS Number: 111524-95-9 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (Neutralization titration) | min. 97.0 % |
Specific rotation [a]20/D | -88.0 to -92.0 deg (C=1, MeOH) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...