Stampa…

9,9-Bis(3,4-dicarboxyphenyl)fluorene Dianhydride ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: 5,5'-(9H-Fluorene-9,9-diyl)bis(2-benzofuran-1,3-dione), BPAF, 5,5'-(9H-Fluorene-9,9-diyl)bis(isobenzofuran-1,3-dione)

F1192-25G F1192-5G
TCIAF1192-25GEA 277 EUR
TCIAF1192-25G TCIAF1192-5G
9,9-Bis(3,4-dicarboxyphenyl)fluorene Dianhydride ≥98.0% (by GC)
9,9-Bis(3,4-dicarboxyphenyl)fluorene Dianhydride
Formula: C₂₉H₁₄O₆
Peso molecolare: 458,43 g/mol
MDL Number: MFCD30342552
CAS Number: 135876-30-1

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Morpholine Method) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR