Stampa…

3-Fluoroazetidine hydrochloride ≥98.0% (by total nitrogen and titration analysis)

Fornitore: TCI
Attenzione

Synonyms: 3-Fluoroazetidine HCl

F1186-200MG F1186-1G
TCIAF1186-200MGEA 24 EUR
TCIAF1186-200MG TCIAF1186-1G
3-Fluoroazetidine hydrochloride ≥98.0% (by total nitrogen and titration analysis)
3-Fluoroazetidine hydrochloride
Formula: C3H7ClFN
Peso molecolare: 111.55 g/mol
MDL Number: MFCD05663715
CAS Number: 617718-46-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=27bc72e802a3f6f6a44c7ec5fe3e03d8daf18039&vid=27bc72e802a3f6f6a44c7ec5fe3e03d8daf18039&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Melting point 128.0 to 132.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR