Stampa…

2-Fluoro-9H-carbazole ≥98.0% (by GC)

Fornitore: TCI
Attenzione

F1182-200MG F1182-1G
TCIAF1182-200MGEA 76 EUR
TCIAF1182-200MG TCIAF1182-1G
2-Fluoro-9H-carbazole ≥98.0% (by GC)
2-Fluoro-9H-carbazole
Formula: C₁₂H₈FN
MDL Number: MFCD09033508
CAS Number: 391-53-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=16c45b1824049c360f563c71b2e17acd71938e12&vid=16c45b1824049c360f563c71b2e17acd71938e12&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 224.0 to 228.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR