6-Fluoro-o-anisaldehyde ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
2-Fluoro-6-mehoxybenzaldehyde, 2-Fluoro-6-methoxybenzaldehyde, 6-Fluoro-2-anisaldehyde
F1175-1G
F1175-5G
TCIAF1175-1GEA
25
EUR
TCIAF1175-1G
TCIAF1175-5G
6-Fluoro-o-anisaldehyde ≥98.0% (by GC)
6-Fluoro-o-anisaldehyde
Formula:
C₈H₇FO₂ Peso molecolare: 154,14 g/mol |
MDL Number:
MFCD01090998 CAS Number: 146137-74-8 |
Risultati del test di specificazione
Appearance | White to Yellow to Orange powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 59.0 to 63.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...