4-Fluoro-3-nitrobenzenesulphonamide ≥98.0% (by HPLC, total nitrogen)
Fornitore: TCI EUROPE
|
Attenzione
|
F1172-1G
F1172-5G
TCIAF1172-1GEA
103
EUR
TCIAF1172-1G
TCIAF1172-5G
4-Fluoro-3-nitrobenzenesulphonamide ≥98.0% (by HPLC, total nitrogen)
4-Fluoro-3-nitrobenzenesulphonamide
Formula:
C₆H₅FN₂O₄S Peso molecolare: 220,18 g/mol |
MDL Number:
MFCD08703185 CAS Number: 406233-31-6 |
Risultati del test di specificazione
Appearance | White to Yellow to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 137.0 to 141.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...