Fulvestrant ≥98.0% (by HPLC)
Fornitore: TCI
|
Pericolo
|
Formula:
C₂₃H₄₇F₅O₃S |
MDL Number:
MFCD00903953 CAS Number: 129453-61-8 UN: 3077 ADR: 9,III |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Specific rotation [a]25 | +108 to +115 deg (365nm, C=2, methanol) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...