Stampa…

(5-Fluoro-2-nitrophenyl)acetic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 5-Fluoro-2-nitrophenylacetic acid

F1140-1G F1140-5G
TCIAF1140-1GEA 43 EUR
TCIAF1140-1G TCIAF1140-5G
(5-Fluoro-2-nitrophenyl)acetic acid ≥98.0% (by GC, titration analysis)
(5-Fluoro-2-nitrophenyl)acetic acid
Formula: C₈H₆FNO₄
Peso molecolare: 199,14 g/mol
MDL Number: MFCD07787601
CAS Number: 29640-98-0

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 153.0 to 157.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR