Stampa…

5-Formylthiophene-2-carbonitrile ≥98.0% (by GC)

Fornitore: TCI
Pericolo

F1133-1G F1133-5G
TCIAF1133-1GEA 75 EUR
TCIAF1133-1G TCIAF1133-5G
5-Formylthiophene-2-carbonitrile ≥98.0% (by GC)
5-Formylthiophene-2-carbonitrile
Formula: C₆H₃NOS
MDL Number: MFCD09037804
CAS Number: 21512-16-3
UN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c47b101d32da33ebe431517308421ee18b61b246&vid=c47b101d32da33ebe431517308421ee18b61b246&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 91.0 to 95.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR